Learn More
Agilent BioTek ELx405 Select Deep Well Microplate Washer
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates.
Brand: Agilent BioTek ELX405CV-SN
Description
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. The ELx405 Select is available in several configurations to address a wide variety of plate washing applications, including ELISA and cell-based assays.
Maintenance is easy with the available patented Ultrasonic Advantage that automates and cleans manifold tubes to remove built-up salt and protein.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Certifications
CE and ETL marked
Specifications
CE and TUV marked. RoHS compliant. | |
≤2 μL/well | |
Microplate Washer | |
1 to 10 | |
One positive displacement pump | |
50/60 Hz | |
59°F to 86°F | |
96-/384-well manifold; Deep well; Automatic 4-Buffer Switching; Optimized cell washing for loosely adherent monolayers | |
35.6 cm | |
25.4 cm | |
43.2 cm | |
100/240 V |
≤3% CV | |
Programmable in minutes and seconds, up to 60 min. | |
13.5 kg | |
Backlit LCD | |
10 to 80%, noncondensing | |
ELX405UCWVD | |
15°C to 30°C | |
14 in. | |
10 in. | |
17 in. | |
30 lb. | |
<8 A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.