missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek ELx808 Absorbance Microplate Reader

Store up to 55 user-programmable protocols. ELx808 Absorbance Microplate Readers read 96-well microplates and provide precise, accurate, and repeatable results.

Specifications

Accuracy >lt;1% at 2.5 O.D., >lt;2% at 3.5 O.D.
Includes Filters, tungsten halogen light source, parallel printer cable, null modem serial cable and Gen5 reader control software
Linearity >lt;1% at 2.5 O.D. (typical)
Weight (English) 30 lb.
Weight (Metric) 13.6 kg
View More Specs

Description

Description

  • Readers can perform multiple assays per plate
  • On-board diagnostic self-test may be used with an optional calibration test plate to easily confirm and document performance
  • Kinetic rate, R2, or onset time is reported without a PC
  • Linear well scanning is ideal for agglutination assays
  • Staggered optical design eliminates channel-to-channel crosstalk
  • Reference channel rolls optical signal for uniform channel-to-channel signal
  • Curve-fitting options include linear, 4-P, 2-P (logit/log), cubic, quadratic, cubic spline, and point-to-point
  • Measurements can be read in intervals as short as six seconds
  • Additional data analysis options include cutoff formulas, endpoint, and kinetic analysis Supplied with 4-Zone Temperature Control
  • Extended dynamic range

Model ELx808

  • Data analysis options include data transformation formulas, assay validation, and control validation
  • Optional 4-Zone Temperature Control improves reproducibility in enzyme kinetic reactions

Order Info

Optional Gen5 Data Analysis Software (Mfr. No. GEN5) provides simple, fast data collection, calculation, exporting, and reporting. Or select Gen5 Secure software to meet requirements of 21 CFR Part 11.

Certifications

CE marked

Specifications

Specifications

>lt;1% at 2.5 O.D., >lt;2% at 3.5 O.D.
>lt;1% at 2.5 O.D. (typical)
13.6 kg
Filters, tungsten halogen light source, parallel printer cable, null modem serial cable and Gen5 reader control software
30 lb.
Videos
SDS
Documents

Documents

Product Certifications