missing translation for 'onlineSavingsMsg'
Learn More

Embelin, Thermo Scientific Chemicals

Product Code. 15406189
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

An inhibitor of XIAP (X-linked inhibitor-of-apoptosis protein) that contains antitumor and anti-inflammatory activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An inhibitor of XIAP (X-linked inhibitor-of-apoptosis protein) that contains antitumor and anti-inflammatory activity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 550-24-3
Molecular Formula C17H26O4
Molecular Weight (g/mol) 294.39
MDL Number MFCD00016369
InChI Key IRSFLDGTOHBADP-UHFFFAOYSA-N
Synonym embelin, embelic acid, emberine, 2,5-dihydroxy-3-undecyl-1,4-benzoquinone, 2,5-dihydroxy-3-undecyl-p-benzoquinone, 2,5-dihydroxy-3-undecyl-2,5-cyclohexadiene-1,4-dione, unii-shc6u8f5er, embelin, embelia ribes, xiap inhibitor, embelin, shc6u8f5er
PubChem CID 3218
ChEBI CHEBI:4778
IUPAC Name 2,5-dihydroxy-3-undecylcyclohexa-2,5-diene-1,4-dione
SMILES CCCCCCCCCCCC1=C(O)C(=O)C=C(O)C1=O

Specifications

Color Orange
Quantity 50 mg
Sensitivity Light sensitive
Merck Index 14,3555
Formula Weight 294.4
Physical Form Solid
Chemical Name or Material Embelin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'