missing translation for 'onlineSavingsMsg'
Learn More

Merck Hazardous Waste Filtration System

Separates solid and liquid phases of waste samples

Brand:  Merck YT30142HW

13132.20 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 72

Additional Details : Weight : 0.10000kg

Product Code. 10054840

  • £14090.00 / Each
Estimated Shipment: 03-09-2024
to see stock.
Add to basket

Description

Description

If your waste samples contain non-volatile or semi-volatile substances or metals, use this Hazardous Waste Filtration System for saparation, extratction, and filtration. Developed in collaboration with the US Environmental Protection Agency (EPA), the system is designed specifically for the US EPA Extraction Procedure (EP) Toxicity Test and Toxicity Characteristic Leaching Procedure (TCLP). The device separates solid and liquid phases of waste samples for analysis. Features & Benefits:•Contains interior holder with PTFE coating to prevent heavy metal contamination •Allows easy introduction of bulky samples through wide entry port and removable top plate•Disassembles for easy cleaningApplications:US EPA EP Toxicity Test and Toxicity Characteristic Leaching ProcedureYT31042HW Hazardous Waste Filtration System, 142 mmReplacement Parts:YY2214257 Hand KnobYY2004055 1-1/2 in. TC Silicone Gasket YY2004045 Clamp 1-1/2 in. TCYY2214265 O-ring (2-251), siliconeYY2214253 O-ring (2-251), PTFEYT3014257 Underdrain Support, PTFE-coatedYY2214251 Legs with Caps, 3/16 in. wrenchYY2214252 Handwheel Wrench for 142 mm HolderAccessories:YY2004057 Gasket, 1-1/2 in., PTFE-coated fluoroelastomerXX6700035 Tubing for Pressure Use, 1/2 in. I.D.Membrane Filters:AP4014250 Glass Fiber filter without Binder, AP40, 142 mmAPFF14250 Glass Fiber Filter without Binder, APFF, 142 mmYT310RAHW Rotary Agitator
Specifications

Specifications

Not Autoclavable
316 Stainless Steel; Aluminum Legs; Molded PP Handwheels; PTFE Seal
Environmental Test Kits or Supplies
42.2 cm
7.25 in.
Hazardous Waste System
Non-sterile
16.6 in.
7.25 in.
18.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers