missing translation for 'onlineSavingsMsg'
Learn More

Merck Hydrosol Stainless Steel Filter Holder and Accessories

For particulate or biological contamination analysis via vacuum filtration

Brand:  Merck XX2004720

1388.14 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 72

Additional Details : Weight : 0.10000kg

Product Code. 10519412

  • £1585.00 / Each
Estimated Shipment: 24-09-2024
to see stock.
Add to basket

Includes: Grounding set for use with flammable liquids; stainless-steel funnel, base, and filter support screen; anodized aluminum locking ring; nylon lockwheels; PTFE resin gaskets; silicone stopper.

Description

Description

The Hydrosol filter holder is designed for the filtration of flammable liquids. Unlike other filter holders, the Hydrosol filter holder includes a grounding unit to prevent sparking from static discharge, which can be a safety hazard. Applications:Vacuum Filtration of Flammable Liquids XX2004720 Hydrosol Filter Holder, 47 mm, stainless steelReplacement PartsXX2004704 Funnel and Locking Ring Assembly, stainless steelXX2004708 Support Screen, 47 mm, stainless steelXX2004702 Base with Screen, gasket and supportXX2004703 Gasket, PTFE XX2004718 No. 8 Stopper, 9.5 mm (3/8 in.) hole, siliconeXX20047RK Replacement Parts Kit for Hydrosol Filter Holder AccessoriesXX1004705 Vacuum Filtering Flask, 1 LXX1004744 Vacuum Filtering Flask, 4LXX7100004 Tubing 3/16 in. I.D. x 140 cm, siliconeXX6200006P Filter Forceps, blunt end, stainless steelXX2604735 3-place Manifold, 47 mm, PVCXX2504735 3-place Manifold, 47 mm, stainless steelXX2504700 6-place Manifold, 47 mm, stainless steelWP6211560 High Output Pump, 115 V/60 HzWP6222050 High Output Pump, 220 V/50 HzWP6210060 High Output Pump, 100 V/50-60 Hz
Specifications

Specifications

Not Autoclavable
Non-sterile
22.9 cm
Stainless Steel
Laboratory Filter Holders or Cyclones
11.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers