missing translation for 'onlineSavingsMsg'
Learn More

Merck Protein G Reagent Kit

Reagents for Amicon Pro Affinity Concentration Kit Protein G with Amicon Ultra–0.5 Device

Brand:  Merck ACR5000PG

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 15534665

  • £375.00 / 1 set
Estimated Shipment: 15-11-2024
to see stock.
Add to basket

Description

Description

For use with the Amicon Pro Affinity Conc: 3 ml Protein G resin, 2 X 66ml Bind/Wash Buffer, 12 ml Elution buffer, 2.5 ml Neutralization buffer
Specifications

Specifications

Single-use
Non-sterile
Regenerated Cellulose (RC)
1Pack
15 mL
Amicon Pro Affinity
Antibody purification
3 mL Protein G resin, 2 x 66 mL Bind/Wash Buffer, 12 mL Elution buffer, 2.5 mL Neutralization buffer
Centrifugal Filters
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers