missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Pro Purification System with Ultra-0.5 Device, 10 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
Brand: Merck ACS501024
This item is not returnable.
View return policy
Description
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.Specifications
Single-use | |
Non-sterile | |
24 Amicon Ultra NMWCO 10 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap | |
Centrifugal Filters | |
12 cm |
10 kDa | |
Regenerated Cellulose (RC) | |
24Pack | |
10 mL |