missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Pro Purification System with Ultra-0.5 Device, 3 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
£241.00 - £348.00
Specifications
Disposable | Single-use |
---|---|
Sterility | Non-sterile |
Housing Material | Regenerated Cellulose (RC) |
Type | Centrifugal Filters |
Volume (Metric) | 15 mL |
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
15339605
|
Merck
ACS500312 |
12 Amicon Ultra NMWCO 3 kDa filters, 12 Exchange devices, and 12 Collection tubes with cap | 12Pack | N/A | |||||
15207956
|
Merck
ACS500324 |
24 Amicon Ultra NMWCO 3 kDa filters, 24 Exchange devices, and 24 Collection tubes with cap | 24Pack |
£348.00
Each |
Estimated Shipment: 08-11-2024
Log in to see stock. |
||||
Description
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.Specifications
Single-use | |
Regenerated Cellulose (RC) | |
15 mL |
Non-sterile | |
Centrifugal Filters | |
12 cm |