missing translation for 'onlineSavingsMsg'
Learn More

Merck Amicon™ Ultra-2.0 Centrifugal Filter Units

Reverse spin capability provides consistent recoveries

Brand:  Merck UFC200324

156.41 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 71

Additional Details : Weight : 0.18900kg

 View more versions of this product

Product Code. 11582134

  • £246.00 / Pack of 24
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

  • Concentrate 2mL down to 15 – 70μL
  • True engineered dead stop provides consistent recovery without spinning to dryness
  • Reverse spin aids in sample recovery, especially with small, dilute samples
  • Vertical membrane design aligns with the filtrate for less clogging and faster filtering
  • Ultracel low-binding membrane combines with the innovative vertical housing to provide fast sample processing without sacrificing recovery
  • Fast processing, with 10- to 30-minute spin times

Applications:

  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC
  • Desalting and buffer exchange

Specifications

Specifications

Regenerated Cellulose
1 cm2
Two microcentrifuge tubes
24/Pk.
0.196 in.
3 kDa
Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC: Desalting and buffer exchange
14,000xG for concentration spin; 1000xG for reverse spin
2.0 mL
4.99 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers