Learn More
Merck Millipore BioPak™ Disposable Ultrafilter
Nuclease-free, pyrogen-free water
Brand:  Merck Millipore CDUFBI001
Additional Details : Weight : 0.20700kg
Product Code. 12780556
  • £490.00 / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
Nuclease-free, pyrogen-free water
  • Guaranteed nuclease-free and pyrogen-free performance for three months, regardless of volumes used
  • Disposable filter eliminates the need for regular, lengthy sanitization procesures and hazardous chemicals
  • Fits all lab-scale EMD Millipore Ultrapure water systems: Simplicity portable water systems, Synergy™ ultrapure water systems, Direct-Q™ water systems, and all generations of Milli-Q™ water systems
  • Preserves water quality as well as performance—no loss in system flowrate
  • Cost-effective upgrade for your existing EMD Millipore ultrapure water system
Applications:
Biological techniques requiring nuclease-free and/or pyrogen-free water
Replacement of molecular biology-grade and cell culture-grade bottled water with similar performance
Advanced life science research
Specifications
Specifications
Ultrafiltration
EMD Millipore™ Ultrapure™, Synergy™ Ultrapure™, Direct-Q™ and all generations of Milli-Q™ water systems
Ultrafiltration Cartridge
Production of pyrogen-free and RNase-free water
1 Pack