Learn More
Merck Millipore EZ-Pak™ Membranes
Used for general microbiological analysis. EZ-Pak™ Membranes are sterile, ready-to-use membranes that eliminate contamination risk and save time.
Description
Used for general microbiological analysis. EZ-Pak™ Membranes are sterile, ready-to-use membranes that eliminate contamination risk and save time.
Press lever to uncover half of membrane surface for easy retrieval with forceps; other half remains in protective packaging. Unused membranes remain safely packed in pleated band.
- Mixed Cellulose Ester
- Sterile and grid surface
- Used for general microbiological analysis
- Certified to meet standard methods for the examination of Water and wastewater requirements
Order Info
Membranes are individually wrapped and come in pleated-band formatted cartridges of 150. Each pack includes four cartridges (600 membranes in all). Membrane catalog number, lot number, sterilization date, pore size, and sequential number from 1 to 150 for each membrane cartridge are printed on clear, protective cover.
Compliance
Meet guidelines for Standard Methods for the Examination of Water and Wastewater.
Safety and Handling
- 0.8µm 47mm white gridded EZ-pak filter membrane made of mixed ester cellulose
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use powder to extinguish.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose, octylphenol polyethoxyethanol