Learn More
Merck Filter Forceps
Handle membrane filters without damage
Brand: Merck XX6200006P
66.43 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : 72
Additional Details : Weight : 0.03060kg
Description
- Made from highly polished stainless steel with beveled, unserrated tips
- Sterilize by autoclaving or flaming
- Pack of 3; contains blue, pink and green
Specifications
Beveled, Unserrated | |
Stainless Steel | |
Forceps, Filter | |
3/Pack |