missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MQuant™ pH Test Strips and Indicator Papers
Measure pH onsite without instruments or extensive preparation
£220.00 - £277.00
Specifications
For Use With (Application) | Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination |
---|---|
Storage Requirements | 10°C to 25°C |
Description
Fit into a pocket and provide easy, rapid results—suited to all media in environmental analysis and industrial in-process controls.
- Differentiated color gradations for a more precise evaluation
- Traditional indicator paper or MQuant™ nonbleeding universal strips available
- High-quality papers impregnated with indicator or mixed-indicator solutions, cut to size and dried
- Supplied in a roll format that protects strips against moisture, light or ambient gases for longer storage life
- Special indicator dyes covalently bonded to reagent papers
- Indicator does not bleed, so strips will not contaminate the medium, even if left for long periods of time
- Can be used for three to five years when stored at 10 to 25°C in a light-protected area
- Reseal quickly after each use
- Tests checked and calibrated using certified buffer solutions
- Buffer solutions are traceable to primary reference materials from NIST™ and PTB
Order Info
Packaging: indicator papers come in lengths of 4.8m in roll dispenser; MQuant™ special indicator pH strips come as strips packed in boxes of 100 tests each.
Specifications
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination | |
10°C to 25°C |
Safety and Handling
missing translation for 'shelfLife' : 3 to 5 Years