missing translation for 'onlineSavingsMsg'
Learn More

Merck Microcon-10kDa Centrifugal Filter Unit with Ultracel-10 membrane

Provides efficient concentration, desalting, or buffer exchange of aqueous biological samples.

Brand:  Merck MRCPRT010

541.41 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.28260kg

Product Code. 13165889

  • £693.50 / Pack of 100
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes. With the low-binding Ultracel™ membrane, Microcon™ filters offer:•Typical recoveries of greater than 95%, even for dilute solutions •Reverse spin to maximize recovery, even in the smallest samples •Convenient storage of filtrate or concentrated sample in standard microfuge tube •Concentration factors up to 100X Features:•High recovery for small volumes with reverse spin (concentration factor less than 20X) •Low-binding Ultracel™ membrane •Fast processing
Specifications

Specifications

Green
Ultracel-10
Regenerated Cellulose (RC)
10 mL
Single-use
Non-sterile
Centrifugal Filters
4.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers