Learn More
Merck Millicell™ Cell Culture Insert Plates
Designed to support cell growth and differentiation of endothelial and epithelial cell lines
Brand:  Merck PSHT004R1
 View more versions of this product
Product Code. 11770373
  • £694.00 / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For easier pipetting and isolateral access
  • Can be used manually or with automated cell seeding, feeding and washing systems
  • Optimized to grow and sustain high-integrity cell monolayers
  • Membrane plates have feet to elevate the plate above the work surface when disassembled from the feeder tray to reduce the risk of monolayer contamination
  • Tear-drop shaped receiver wells eliminate air bubbles as plates are assembled
  • Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane
Type and number of components vary by model:
PSHT004R1 Includes
  • One 96-Well Cell Culture Plate
  • One Feeder Tray
  • One 96-Well Receiver Tray
  • One Lid
PSHT004R5 Includes
  • Five 96-Well Culture Plates
  • Five Single-Well Feeder Trays
  • Five Lids
PSHT004S5 Includes
  • Five 96-Well Cell Culture Plates
  • Ten 96-Well Receiver Trays
  • Five Lids
  • Cell Attachment
  • Cell Culture
  • Cell Differentiation
  • Drug Transport
  • Permeability
  • Cell-Based Assays
  • Caco-2 Cell Culture
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PCF
Single-well feeder tray, 96-well receiver tray and lid
Sterile
0.4 μm
Cell Attachment, Cell Culture, Cell Differentiation, Drug Transport
96