missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millicell™ Cell Culture Insert Plates
Designed to support cell growth and differentiation of endothelial and epithelial cell lines
£650.00 - £3145.00
Specifications
Material | PCF |
---|---|
Pore Size | 0.4 μm |
For Use With (Application) | Cell Attachment, Cell Culture, Cell Differentiation, Drug Transport |
Sterility | Sterile |
No. of Wells | 96 |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11700393
|
Merck
PSHT004S5 |
Single-well feeder tray and lid |
£3145.00
Pack of 5 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
11770373
|
Merck
PSHT004R1 |
Single-well feeder tray, 96-well receiver tray and lid |
£650.00
Each |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
11790383
|
Merck
PSHT004R5 |
96-well receiver tray and lid |
£3115.00
Pack of 5 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
Description
- For easier pipetting and isolateral access
- Can be used manually or with automated cell seeding, feeding and washing systems
- Optimized to grow and sustain high-integrity cell monolayers
- Membrane plates have feet to elevate the plate above the work surface when disassembled from the feeder tray to reduce the risk of monolayer contamination
- Tear-drop shaped receiver wells eliminate air bubbles as plates are assembled
- Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane
Type and number of components vary by model:
PSHT004R1 Includes
- One 96-Well Cell Culture Plate
- One Feeder Tray
- One 96-Well Receiver Tray
- One Lid
- Five 96-Well Culture Plates
- Five Single-Well Feeder Trays
- Five Lids
- Five 96-Well Cell Culture Plates
- Ten 96-Well Receiver Trays
- Five Lids
- Cell Attachment
- Cell Culture
- Cell Differentiation
- Drug Transport
- Permeability
- Cell-Based Assays
- Caco-2 Cell Culture
Specifications
PCF | |
Cell Attachment, Cell Culture, Cell Differentiation, Drug Transport | |
96 |
0.4 μm | |
Sterile |