Learn More
Merck Millicell™ EZ Slides
Culture, fix, stain, and view cells all in one device
Brand: Merck PEZGS0496
Code : 71
Additional Details : Weight : 0.10000kg
Description
- Polypropylene wells are resistant to most fixatives—cells may be fixed and stained with wells still attached
- Breakable tabs for easy well removal—no tools required to remove wells from slide
- No adhesive used to attach wells to slide
- No leaky wells
- Raised corner ridges on lid facilitate stacking of multiple units in the incubator
- Noncytotoxic and nonpyrogenic
- Sterile
- Slide holder (available separately) protects slide from damage
Specifications
EZ Slide | |
Clear | |
1.7 cm2/well | |
Glass | |
Simplify your cell analysis by using the Millicell EZ SLIDE glass slide to culture, fix, stain and view your sample all in one device |
4 | |
Polypropylene | |
Polystyrene | |
0.5 to 1.7 mL/Well | |
Sterile |