Learn More
Merck MultiScreen™ 96-Well Assay Plates
Provide tools to perform high-throughput studies of ADME processes—the investigation of the adsorption, distribution, metabolism, and excretion of potential drugs
Brand:  Merck MAIPN4550
Code : 73
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10741801
  • £1575.00 / Pack of 50
Estimated Shipment: 14-08-2024
to see stock.
Add to basket
Description
Description
Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.
  • Use with a vacuum manifold, automated liquid-handling system, or microplate scintillation counter—entire assays can be performed on one plate
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
  • Also ensures quantitative filtrate transfer to a receiver plate placed in bottom of manifold
Choice of filter materials
  • For high protein binding applications, use plates with Immobilon™-P PVDF or mixed cellulose esters For high nucleic acid binding, use plates with mixed cellulose esters, glass fiber, or positively charged DEAE membranes For charged biomolecule separations, choose from positively charged DEAE or negatively charged phosphocellulose membranes
  • For low biomolecule binding, use plates with hydrophilic Durapore™ PVDF, Biopore™ PTFE, or polycarbonate membranes
  • For assays that require solvent resistance, use plates with hydrophilic PTFE membranes
Each microporous membrane interacts distinctly with different proteins or biomolecules; therefore, it may be necessary to test different membrane types for each assay.
†Includes lid, filter plate, and receiver plate.
‡Includes extended centrifugal cover, filter plate, and receiver plate.
††Consists of a 96-place holder and a 96-well receiver plate preloaded with Microcon YM-10 filter units.
Specifications
Specifications
Standard
8 x 12
Clear
ABS Plastic
Non-sterile
50/Pk.
Hydrophobic PVDF
0.45 μm
MultiScreen-IP Filter Plate
96
Avidin biotin linkages, DNA-binding proteins, protein binding, PAMPA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products