missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
£464.00 - £1745.00
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Pore Size | 0.45 μm |
No. of Wells | 96 |
For Use With (Application) | ELISPOT assays |
Product Code | Brand | Description | Color | Material | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Color | Material | Sterility | Price | Quantity & Availability | ||
10258100
|
Merck
MAHAS4510 |
MultiScreen™-HA with MCE membrane | Clear | Styrene | Sterile |
£482.00
Pack of 10 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||
10146580
|
Merck
MAIPS4510 |
MultiScreen™-IP plate with Immobilon™-P membrane without underdrain | Clear | ABS Plastic | Sterile |
£504.00
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
10307473
|
Merck
MSIPN4W50 |
MultiScreen-IP Filter Plate with Immobilon™-P membrane | White | ABS Plastic | Non-sterile |
£1745.00
Pack of 50 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
||
10402362
|
Merck
MSIPS4W10 |
MultiScreen™HTS-IP plate with Immobilon™-P membrane | White | Acrylic | Sterile |
£464.00
Pack of 10 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
10532844
|
Merck
S2EM004M99 |
MultiScreen-IP Filter Plate with Immobilon™-P membrane | White | Acrylic | Sterile |
£484.00
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
10659303
|
Merck
MAIPSWU10 |
MultiScreen™HTS plate with Immobilon™-P membrane without underdrain | White | Acrylic | Sterile |
£636.00
Pack of 10 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
- Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
Specifications
Standard | |
0.45 μm | |
ELISPOT assays |
8 x 12 | |
96 |