Learn More
Merck MultiScreen™ 96-Well Assay System Accessory: Feeder/Transport Trays
Perform cell growth, feeding, and analysis in one plate
Brand:  Merck MACAC0RS5
Code : 73
Additional Details : Weight : 0.10000kg
Product Code. 10307663
  • £209.00 / Pack of 5
Estimated Shipment: 06-08-2024
to see stock.
Add to basket
Includes: (5) 96-well Feeder Trays; (5) transport trays with lids
Description
Description
  • With Lid
Specifications
Specifications
Standard
8 x 12
Polystyrene
Sterile
Caco-2 cell culture, cell-based assays
With Lid
Clear
96
0.28 cm2
5/Pk.