missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ High Volume Filter Plates

Multiwell solutions for discovery research and sample prep

Brand:  Merck MVHVN4525

Code : NEW

Additional Details : Weight : 3.81000kg

 View more versions of this product

Product Code. 15422263

  • £1470.00 / Pack of 25
Estimated Shipment: 20-11-2024
to see stock.
Add to basket

Description

Description

MultiScreen filter plates meet specific performance criteria for sample preparation methodology requiring low nonspecific binding of protein and drug analytes, solvent compatibility and sample throughput. The MultiScreen High Volume filter plate can process up to 2mL of aqueous and organic solutions, with best-in-class discrete and quantitative filtrate transfer to a custom designed 96-well MultiScreen High Volume collection plate using the MultiScreen hts vacuum manifold. The MultiScreen High Volume filter plate is fully automation-compatible and complies with ANSI Deepwell Plate standards.

MultiScreen High Volume filter plate with Durapore™ membrane

  • For high recovery (low nonspecific binding) of small molecules and proteins, low extractables, and compatibility with aqueous and some organic solutions
  • Applications include: low protein binding assays, low extractables assays, sample cleanup, particulate/aggregate removal (prior to HPLC or UPLC™ analysis), and bead-based assays
MultiScreen High Volume filter plate with 1.2μm Glass Fiber C (GFC) membrane
  • Offers broader chemical compatibility with both aqueous and organic solutions
  • Applications requiring high flow including: lysate clearing, viscous sample filtration, environmental testing, and DNA sample preparation bind-wash-elute applications

Specifications

Specifications

Standard
8 x 12
Opaque
Polypropylene
96
Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase precipitation assays, bead, resin based assays
1.27 cm
4.7 cm
Hydrophilic PVDF
0.45 μm
MultiScreen High Volume HV Plate
2 mL
Non-sterile
0.5 x 3.37 x 1.85 in. (1.27 x 8.56 x 4.7 cm)
8.56 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers