missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ PH Plate
Reliable tools for the advancement of drug discovery and life science research
Brand: Merck MAPHN0B50
This item is not returnable.
View return policy
Description
- Negatively charged phosphocellulose
- Low protein/nucleic acid binding
- Opaque
- Non-Sterile
Protein kinase assays with peptide substrates, enzyme assays, binds positive charge
Specifications
Opaque | |
Phosphocellulose | |
Non-sterile | |
0.28 cm2 | |
0.001 to 0.008 oz. | |
5.03 x 3.3 x 0.56 in. (12.8 x 8.55 x 1.44cm) | |
12.8 cm | |
8.55 cm | |
1.44 cm |
Barex/TiO2 | |
0.65 μm | |
PH | |
Protein kinase assays with peptide substrates, enzyme assay, binds positive charge, reliable tools for the advancement of drug discovery and life science research | |
0.05 to 0.25 mL | |
5.03 in. | |
3.3 in. | |
0.56 in. |