missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ Sealing Tape

Product Code. p-155647 Shop All Merck Products
Click to view available options
Color:
Clear
Opaque
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11758522

Brand: Merck MATAH0P00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Choose from opaque or clear

  • Opaque sealing tape is used to seal 100μL plate bottoms read in TopCount™ instruments
  • Clear sealing tape can be used to seal all manufacturers' 300μL plate tops and plate bottoms for MicroBeta™ readers
  • Precut to size

TRUSTED_SUSTAINABILITY

Specifications

Color Opaque
Sterility Non-sterile
No. per Pack 100
For Use With (Application) Used to seal 100μL plate bottoms read in TopCount™ instruments
Product Type Sealing Tape
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.