missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ Solvinert Filter Plates
Optimized for drug discovery applications. MultiScreen™ Solvinert Filter Plates are solvent compatible and feature a unique one-piece design. 96-well plates are available in deep well (2mL) format (with or without pre-filter) and standard (500μL) format, with hydrophobic or hydrophilic membranes.
£492.00 - £2240.00
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Pore Size | 0.45 μm |
Color | Clear |
Material | Polyolefin Copolymer |
Product Code | Brand | Membrane | Height (Metric) | No. per Pack | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Membrane | Height (Metric) | No. per Pack | Price | Quantity & Availability | |||
10184123
|
Merck
MDRPN0410 |
Hydrophobic PTFE | 4.07 cm | 10/Pk. |
£965.00
Pack of 10 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||
10307083
|
Merck
MDRLN0410 |
Hydrophilic PTFE | 4.07 cm | 10/Pk. |
£950.00
Pack of 10 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||
10245943
|
Merck
MSRLN0410 |
Hydrophilic PTFE | 1.46 cm | 10/Pk. |
£492.00
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10478603
|
Merck
MSRLN0450 |
Hydrophilic PTFE | 1.46 cm | 50 |
£2085.00
Pack of 50 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10031863
|
Merck
MSRPN0410 |
Hydrophobic PTFE | 1.44 cm | 10/Pk. |
£492.00
Pack of 10 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||
10114323
|
Merck
MSRPN0450 |
Hydrophobic PTFE | 1.46 cm | 50 |
£2240.00
Pack of 50 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||
Description
- Clear polyolefin base plate offers broad solvent compatibility
- One-piece design eliminates leaking and allows for extended incubations
- Low binding and low extractables
- Accommodate aqueous and nonaqueous samples with chemically resistant hydrophilic or hydrophobic membranes
- Polytetrafluoroethylene membranes feature 45μL pores
- Automation compatible
- Available in 96-well deep well (2mL) format (with or without pre-filter) and standard (500μL) format
Specifications
Standard | |
0.45 μm | |
Polyolefin Copolymer | |
Non-sterile | |
1.28 cm |
8 x 12 | |
Clear | |
96 | |
Solid phase synthesis and cleanup, in plate protein precipitation, sample recovery following in-plate compound cleavage, peptide synthesis | |
8.55 cm |