missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ Solvinert Filter Plates

Optimized for drug discovery applications. MultiScreen™ Solvinert Filter Plates are solvent compatible and feature a unique one-piece design. 96-well plates are available in deep well (2mL) format (with or without pre-filter) and stand

Brand:  Merck MDRPNP410

Code : 73

Additional Details : Weight : 0.10000kg

Product Code. 10504574

  • £1085.00 / Pack of 10
Estimated Shipment: 29-08-2024
to see stock.
Add to basket

Description

Description

  • Clear polyolefin base plate offers broad solvent compatibility
  • One-piece design eliminates leaking and allows for extended incubations
  • Low binding and low extractables
  • Accommodate aqueous and nonaqueous samples with chemically resistant hydrophilic or hydrophobic membranes
  • Polytetrafluoroethylene membranes feature 45μL pores
  • Automation compatible
  • Available in 96-well deep well (2mL) format (with or without pre-filter) and standard (500μL) format
Specifications

Specifications

Standard
8 x 12
Clear
96
0.28 cm2
10/Pk.
1.28 cm
4.07 cm
Hydrophobic PTFE
0.45 μm
Polyolefin Copolymer
Non-sterile
Solid phase synthesis and cleanup, in plate protein precipitation, sample recovery following in-plate compound cleavage, peptide synthesis
10/Pk.
8.55 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers