Learn More
Merck MultiScreen™HTS 8-Well Strips with Immobilon™-P PVDF Membrane
8-well strips enable processing of less than 96 samples at one time
Brand: Merck M8IPFRAME
Includes: Twelve 8-well strips in a support frame. Additional support frames also available separately.
Description
- For ELISpot and protein binding assays
Specifications
Strip Plate | |
Clear | |
8-well strips | |
8 |
1 x 8 | |
Microplate Strip Support Frame | |
ABS Plastic | |
Sterile |
Product Content Correction
Product Title
Merck MultiScreen™HTS 8-Well Strips with Immobilon™-P PVDF Membrane > Microplate Strip Support Frame
Spot an opportunity for improvement?Share a Content Correction