missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreenHTS 96-Well Filter Plates
Brand: Merck MSIPN4550
Code : 73
Additional Details : Weight : 1.00000kg
Specifications
ANSI, SBS | |
Hydrophobic PVDF | |
0.45 μm | |
Clear | |
96 | |
0.28 cm2 | |
50/Pk. |
Standard | |
8 x 12 | |
Low Binding | |
ABS Plastic | |
Non-sterile | |
Protein interaction assays, avidin biotin linkages, DNA binding proteins |