Learn More
Merck PureProteome™ Magnetic Beads
For rapid and reproducible isolation of proteins
Brand:  Merck LSKMAGN04
 View more versions of this product
Product Code. 12541219
  • £1020.00 / Pack of 4
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Custom affinity bead generation for binding to target ligand
  • Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Beads are immobilized in seconds
  • Increased kinetics of bead-protein binding enables shorter incubations
  • Significantly more affordable then competitive magnetic beads
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NHS flexiBind
10 μm
Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions
4 x 0.5 mL
2° to 8°C
Magnetic Beads