Learn More
Merck™ Replacement Receiver Flasks
Provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33mm) or wide-mouth (45mm) bottles
Description
Description
  • Sterile, polystyrene
  • Can be used as receivers for Millipore Steritop™ Vacuum Bottle-Top Filters
  • 45mm neck with screw-top lid
  • Storage of sterilized or clarified fluids
  • Laboratory Filtration