Learn More
Merck Replacement Solutions for Montage™ BAC96 Miniprep Kit and Montage Plasmid Miniprep96 Kit
Suitable for cell resuspension for plasmid & BAC minipreps
Brand: Merck LSKCLS500
Code : NEW
Additional Details : Weight : 0.36800kg Transport : UN number : 1824 Chem class : 8 Pack group : III
Description
The Montage Plasmid MiniprepHTS 96 Kit is a fast, easy-to-use kit for high-purity plasmid or bacterial artificial chromosome (BAC) miniprepsSpecifications
Suitable for Alkaline Lysis of plasmid and BAC preparation | |
Cell Lysis Solution | |
500 mL |