Learn More
Merck Samplicity™ Filtration System Vial Trays
Vacuum filtration system for sample preparation for HPLC
£144.00 - £200.00
Specifications
For Use With (Equipment) | Samplicity G2 Filtration System |
---|---|
Sterility | Non-sterile |
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
12920265
|
Merck
SAMVIALTR |
Vial Trays | Polypropylene |
£200.00
Pack of 2 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||
12541249
|
Merck
SAMWASTTR |
Waste Trays | Recycled Polyester |
£144.00
Pack of 5 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||
Description
Overview of Samplicity™ G2 System- Samplicity™ G2 Filtration System enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
- Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex™ 33 mm filter (Millex™ Filters for the Samplicity™ G2 System) or an integrated funnel and filter (Millex Samplicity™ filter)
- Samplicity™ G2 System has lower ergonomic impact than manual syringe filtration
Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC
Specifications
Samplicity G2 Filtration System | |
Non-sterile |