Learn More
Merck Luminata™ Starter Package
Western blot packages with unbelievable savings! Save time and money by choosing the package that best serves your needs!
Brand:  Merck WBLUM0100
Product Code. 12541289
  • £469.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Samples of all three Luminata Western HRP substrates for antibodies of all affinities
  • Luminata Classico Western HRP Substrate, 100mL (WBLUC0100)
  • Luminata Crescendo Western HRP Substrate, 100mL (WBLUR0100)
  • Luminata Forte Western HRP Substrate, 100mL (WBLUF0100)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western blotting, binding assays, amino acid analysis, N-terminal protein sequencing, dot/slot blotting, glycoprotein visualization, lipopolysaccharide analysis.
Snap i.d.™ Protein Detection System
Luminata Starter Kit
Luminata Classico Western HRP Substrate, 100mL (WBLUC0100)
Luminata Crescendo Western HRP Substrate, 100mL (WBLUR0100)
Luminata Forte Western HRP Substrate, 100mL (WBLUF0100)