Learn More
Merck SteriCap PLUS
For ultrafast, aseptic sterilization of 2 to 10 L using vacuum filtration
Brand: Merck SCGPCAPRE
222.15 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : 73
Additional Details : Weight : 0.93600kg
Description
The Stericap™ PLUS Universal Bottle-Top Filter Device with Millipore Express™ PLUS membrane provides fast vacuum filtration and low protein binding without the need for a prefilter in most cases. It filters quickly, without clogging, even viscous solutions typical of serum-containing media. This non-cytotoxic, non-pyrogenic device can be used to fill any vacuum-rated bottle or other container with an inner neck diameter of 20 to 67 mm. This allows maximum versatility, capacity, speed, and flowthrough efficiency when you need to filter larger volumes into containers with openings of various sizes.
Features & Benefits:
- Millipore Express™ PLUS membrane for fastest flow rates and low protein binding
- Fits any vacuum-rated bottle from 20 to 67 mm in diameter for maximum flexibility
- High volume capacity for larger and less frequent preparation steps
- Easy flow control button so the filter can be easily transferred from bottle to bottle without turning off the vacuum each time
- Free of cytotoxic and pyrogenic contaminants for robust cell culture and sensitive downstream applications
Applications: Sterilization of Aqueous Solutions, Media Preparation, Buffer Preparation
Specifications
Clear | |
Single-use | |
Styrene, Polypropylene, Polyethylene | |
0.22 μm | |
40 cm2 | |
10Pack | |
Universal bottle top | |
45°C | |
150 mL | |
7.6 cm | |
10 cm |
Hose barb | |
Express PLUS | |
PES | |
Sterile | |
To sterilize tissue culture media and additives, protein solutions, virus suspensions, DNA, and other aqueous solutions, to help prevent filter air lock, laboratory filtration | |
Universal bottle top | |
113°F | |
Bottle Top Filter | |
2.99 in. | |
10 cm | |
7.3 cm |