missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for ZHE Hazardous Waste System

Accessories for filter holders for sample preparation.

Brand:  Merck YT3009015

787.71 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10430683

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
ZHE Hazardous Waste filtration system
Non-sterile
ZHE Tight Syringe, 50mL
Borosilicate Glass
ZHE Tight Syringe, 50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers