Learn More
BD Emerald™ Three-Part Syringe
Combines optimum performance and reduced environmental impact. BD™ Emerald™ Three-Part Syringe is a general purpose syringe that contains up to 30% less material than other syringes to minimize waste. Syringes feature a center-mounted Luer connection and no needle.
Brand:  BD 307736
 View more versions of this product
Product Code. 15205093
  • £72.24 / Pack of 100
Edge
Explore more special offers
Description
Description
  • Combines optimum performance and reduced environmental impact
  • Manufactured in a production plant powered by renewable energy
  • Crystal clear plastic allows for easy viewing of the contents
  • Green-colored piston crown and engraved shell pressure for precise and safe dosage
  • Robust piston stop prevents the rod from accidentally being knocked out
  • Rod designed for stable and comfortable grip during use
  • Thumb grip with the structure for secure grip during administration
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE marked, ISO 7886-1 and 549 compliant
General purpose use and pressure applications
0.2 mL
10 mL
Yes
Clear with Green Piston Crown
3-part Syringe
100/Pk.
Centric Luer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Emerald™ Three-Part Syringe > Capacity: 10mL