Learn More
3M™ Emphaze™ Polyimide Film Tape 5413
Made with polyimide film and a silicone adhesive, useful for PCB solder masking and other high temperature applications.
Brand:  3M™ 7100010443
Product Code. 17135034
  • £36.35 / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use 3M™ Polyimide Film Tape 5413 for PCB solder masking and other high temperature applications within a range of 73 to 260°C. This amber colored, 0.07 mm thick tape is made with a polyimide film and a silicone adhesive. It also features a polyethylene tape core instead of cardboard.
  • Silicone adhesive's high temperature performance reduces adhesive transfer, helping to eliminate cleaning
  • Polyimide film provides excellent release, as it doesn't soften and stays dimensionally stable at high temperatures
  • Surfaces are protected due to the tape's flame retardancy, and chemical and radiation resistance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Silicone
Polyimide
36 yd.
0.5 in.
Amber
36 yd. x 0.5 in. (33 m x 1.2 cm)
33 m
1.2 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Emphaze™ Polyimide Film Tape 5413 > Polyimide Film Tape; Adhesive Type: Silicone; Color: Amber; Dimensions (LxW): 36 yd. x 0.50 in. (33 m x 1.2 cm); No. per Pack: 2 Pk.