missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Empty FastPrep™ Tubes
Used in Fastprep system to hold sample for Homogenization. MP Biomedicals™ Empty FastPrep™ Tubes are non-skirted without caps.
Brand: MP Biomedicals™ 115076400
322.77 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
- 2mL capacity
- Non-skirted
- Caps not included
- Available empty for those who wish to fill their own tubes
- For use with FastPrep System
Specifications
10.8 mm | |
Natural | |
46mm |
2 mL | |
Polypropylene | |
500/PK |