missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enalaprilat D5, MedChemExpress
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Enalaprilat D5 (MK-422 D5) is the deuterium labeled Enalaprilat(MK-422), which is an angiotensin-converting enzyme (ACE) inhibitor.
Specifications
Specifications
| Chemical Name or Material | Enalaprilat D5 |
| CAS | 349554-00-3 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C18H19D5N2O5 |
| Color | White |
| Synonym | MK-422 D5 |
| SMILES | C[C@@H](C(N1CCC[C@H]1C(O)=O)=O)N[C@H](C(O)=O)CCC2=C([2H])C([2H])=C([2H])C([2H])=C2[2H] |
| Industry Type | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 353.42 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction