Learn More
Day Impex™ Enamelled Steel Container
Safely store and transport liquid gases. Day Impex™ Enamelled Steel Container is specially designed for laboratory, hospital, and industrial use.
£131.00 - £730.00
Specifications
Material | Enameled Steel |
---|---|
For Use With (Application) | Use for liquid gas or as a cold trap |
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
11817452
|
Day Impex™
200003 |
200 mL | 215mm |
£131.00
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
||||
11837452
|
Day Impex™
200005 |
750 mL | 285mm |
£209.00
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
11827452
|
Day Impex™
200004 |
700 mL | 350mm |
£151.00
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11857452
|
Day Impex™
200007 |
1500 mL | 290mm |
£448.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
11847452
|
Day Impex™
200006 |
1100 mL | 275mm |
£197.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
11867452
|
Day Impex™
200008 |
1500 mL | 280mm |
£190.00
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||||
11796399
|
Day Impex™
200014 |
10 L | 445mm |
£730.00
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
11766399
|
Day Impex™
200011 |
1 L | 228mm |
£236.33
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
11776399
|
Day Impex™
200012 |
2 L | 268mm |
£271.42
Each |
In Stock
Log in to see stock. |
||||
11877452
|
Day Impex™
200009 |
2 L | 330mm |
£198.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11786399
|
Day Impex™
200013 |
4500 mL | 350mm |
£250.00
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||||
11887452
|
Day Impex™
200010 |
3 L | 290mm |
£235.00
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
Description
- Safe for use with all liquid gases and ideal for cold traps
- Attractive blue stove-enamelled steel outer container
Specifications
Enameled Steel | |
Use for liquid gas or as a cold trap |