missing translation for 'onlineSavingsMsg'
Learn More

Enantiomer of Sofosbuvir, MedChemExpress

Product Code. p-200097412
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16481248

Brand: Medchem Express HYI0726

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Enantiomer of Sofosbuvir is an enantiomer of Sofosbuvir, a prescription medicine for the treatment of patients with chronic hepatitis C. There is no biological activity report on enantiomer of Sofosbuvir until now.

Enantiomer of Sofosbuvir is an enantiomer of Sofosbuvir, a prescription medicine for the treatment of patients with chronic hepatitis C. There is no biological activity report on enantiomer of Sofosbuvir until now.

Specifications

Chemical Name or Material Enantiomer of Sofosbuvir
Color White
Purity Grade Notes Research
Molecular Formula C22H29FN3O9P
Quantity 5 mg
SMILES O=[P@](N[C@H](C)C(OC(C)C)=O)(OC[C@H]1[C@H](O)[C@@](F)(C)[C@@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3
Molecular Weight (g/mol) 529.45
Formula Weight 529.45
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.