Learn More
SPEX Certiprep™ Extractor for Microvials
Use this to easily removes the end plug from frozen microvials.
Brand: SPEX Certiprep™ 6758
Code : NEW
Additional Details : Weight : 5.00000kg
Description
This end-plug extractor accessory for the SPEX SamplePrep freezer mill 6757 microvial set is a bench tool for easy removal of end plugs from frozen microvials. Simply place three microvials into the extractor cradle and press down on the extractor handle to remove the end plugs. Sample can be retrieved from the vial while it is still cold. Suction cups under the extractor secure it to the benchtop preventing slippage and allowing for one-handed operation.Specifications
End Plug Extractor | |
Holds up to three Microvals | |
6757 Microvial |