Learn More
Syngene Endeavour Ductless Polypropylene Fume Hood with Sliding Sash
Endeavour ductless polypropylene fume hood with AirSafe automatic safety controller.
Brand: Syngene SYN-72EDFH-E
This item is not returnable.
View return policy
Description
72 in. wide Endeavour ductless polypropylene fume hood with AirSafe automatic safety controller, vertical safety glass sliding sash and vapor proof fluorescent light.
- Hood requires a minimum of four like filters for operation, not included in price.
- 220V AC.
Specifications
Ductless Enclosure | |
Polypropylene, Polycarbonate |