Learn More
Endocan Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547206
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse Endocan is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Endocan Polyclonal antibody specifically detects Endocan in Human samples. It is validated for Neutralization, Western Blot.Specifications
Endocan | |
Polyclonal | |
Unconjugated | |
ESM1 | |
0610042H23Rik; AV004503; endocan; endothelial cell specific molecule 1; endothelial cell-specific molecule 1; ESM1; ESM-1; ESM-1 secretory protein; IGFBP-rp6; PG25; pineal specific PG25 protein | |
Goat | |
Antigen affinity chromatography | |
RUO | |
11082 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9NQ30 | |
ESM1 | |
Mouse myeloma cell line NS0-derived recombinant human Endocan/ESM-1 Trp20-Arg184 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |