Learn More
R&D Systems™ Endothelial Cell Growth Media without VEGF
Formulated to support the expansion of human primary endothelial cells under low-serum, VEGF-free conditions
Brand: R&D Systems™ CCM029
Additional Details : Weight : 0.29100kg
Alert:
When handling biohazardous materials such as human cells, safe laboratory procedures should be followed and protective clothing should be worn. This product is for research use only and is not approved for diagnostic procedures.
Description
- Unique and optimized formulation that promotes endothelial cell growth.
- Free of Vascular Endothelial Growth Factor (VEGF)
- Includes a single supplement featuring key growth factors and small molecules
- 250 mL bottle results in less wasted media
- This media has been tested for its ability to support CD31+, KDR+, and vWF+ human umbilical vein endothelial cells growth in vitro
- This kit contains optimized concentrations of the following reagents to maintain and expand human and mouse endothelial cells in culture:
- Endothelial Cell Growth Base Media (250 mL)
- Endothelial Cell Growth Supplement without VEGF (50X)
- Human FGF basic
- Human LR3 IGF-1
- Human EGF
- Heparin
- Hydrocortisone
- L-Ascorbic Acid
- Fetal Bovine Serum
- FOR LABORATORY RESEARCH USE ONLY. NOT FOR USE IN DIAGNOSTIC PROCEDURES.
Specifications
Store the unopened product at 2 - 8 °C. Do not use past expiration date. | |
Growth Factors | |
250 mL |