missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Endothelial Cell Growth Supplement (50X)
Media supplement for low-serum expansion of primary endothelial cells.
Brand: R&D Systems™ 390599
Additional Details : Weight : 0.29100kg
Includes: Human FGF basic, Human LR3 IGF-1, Human VEGF165, Human EGF, Heparin,Hydrocortisone, L-Ascorbic Acid,Fetal Bovine Serum
Description
- Support expansion of human primary endothelial cells under
low-serum conditions - Supports CD31+, KDR+,vWF+, human umbilical vein, endothelial cell growth in vitro
Storage: Store in the dark at 2°C to 8°C. Do not freeze.
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
Growth Supplement | |
Cell Growth and Differentiation | |
5 mL |
For Research Use Only