Learn More
Brady™ Engraved Replacement Labels for M710, M610, M611
Engraved Plate Replacement Labels for use with Brady™ Printers M611, M610 and M710.
Brand:  Brady™ 312102
 View more versions of this product
Product Code. 17857994
  • £60.60 / Pack of 150
Estimated Shipment: 13-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Engraved Plate Replacement labels for the M610 and M611 Thermal Transfer Portable Printers
  • Parts can also be printed on the M710 Label Printer provided the roll adapter (M71-ADAPT) is used
  • Aggressive adhesive and solvent resistance makes them an ideal replacement for engraved tags
  • Service temperature of -40°C to +100°C
Specifications
Specifications
Permanent Acrylic, Foam-backed
Black
Label
Single-side Printable
Engraved Plate Labels
Data and Telecommunications, Electrical Labeling, General Identification
1 Label
150 Labels
-40°F to +212°F
0.0256 in.
15 x 27 mm
1.06 in.
27 mm
None
Gloss
Polyester
Thermal Transfer
Non-sterile
M610, M710, M611, BMP61, BMP71
-40°C
Rectangle
-40°C to +100°C
0.65 mm
0.59 in.
27 mm