Learn More
Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for the iCAP™ Series ICP-OES Systems
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance plasma torch for iCAP™ Series ICP-OES systems.
POA
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
12794868
View Documents
Thermo Scientific™
842312051841
EMT Duo Torch for iCAP 7000 Series ICP-OES systems
POA
Each
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for Thermo Scientific™ iCAP™ Series ICP-OES systems. The torch clips in and can be removed in just seconds.
Enhanced performance through advanced accessories
We offer dedicated torch designs for radial and axial plasma views, including:
  • EMT Radial Torch for radial-only systems
  • EMT Duo Torch for radial and axial (duo) view systems
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for the iCAP™ Series ICP-OES Systems