Learn More
3M™ Enrichment Pouch
A heavyweight, puncture-resistant pouch, ideal for many sample preparation and enrichment procedures.
Brand:  3M™ 7100062091
Additional Details : CAS Number : 7732-18-5 Weight : 0.00200kg
 View more versions of this product
Product Code. 17223764
  • £134.62 / Pack of 32
Estimated Shipment: 04-09-2024
to see stock.
Add to basket
Description
Description
  • Gamma irradiated, ready-to-use pouch with 225 mL Buffered Peptone Water Broth requires no mixing or sterilizing.
  • Flat bottom, stand-up pouch ensures stability when adding samples.
  • Wire tie holds top open for easy sample addition and closes pouch for transportation.
  • Multi-laminate polyester pouch is puncture resistant.
  • 2 year shelf life from date of manufacture provides long-term readiness.
Specifications
Specifications
225 mL Buffered Peptone Water Broth ISO with Filter
32/Cs.