missing translation for 'onlineSavingsMsg'
Learn More

Entacapone, Tocris Bioscience™

Product Code. 30001556
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent COMT inhibitor; blocks α-synuclein aggregation

Potent catechol O-methyltransferase (COMT) inhibitor (IC50 values are 14.3, 20.1 and 73.3 nM for rat liver soluble COMT, total COMT and membrane-bound COMT respectively). Increases bioavailability of levodopa when given as an adjunct therapy for Parkinson's disease. Inhibits α-synuclein aggregation in an in vitro assay; blocks α-synuclein-induced cell death in PC-12 cells.

Chemical Identifiers

CAS 130929-57-6
Molecular Formula C14H15N3O5
Molecular Weight (g/mol) 305.29
MDL Number MFCD00866580
InChI Key JRURYQJSLYLRLN-BJMVGYQFSA-N
Synonym entacapone, comtan, comtess, entacaponum, entacapona, entacaponum inn-latin, entacapona inn-spanish, entacom, entacapone usan:inn, 2-cyano-n,n-diethyl-3-3,4-dihydroxy-5-nitrophenyl propenamide
PubChem CID 5281081
ChEBI CHEBI:4798
IUPAC Name (2E)-2-cyano-3-(3,4-dihydroxy-5-nitrophenyl)-N,N-diethylprop-2-enamide
SMILES CCN(CC)C(=O)C(=C\C1=CC(O)=C(O)C(=C1)[N+]([O-])=O)\C#N

Specifications

Quantity 10 mg
Formula Weight 305.29
Percent Purity >99%
Chemical Name or Material Entacapone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.