missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Entacapone, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent catechol O-methyltransferase (COMT) inhibitor (IC50 values are 14.3, 20.1 and 73.3 nM for rat liver soluble COMT, total COMT and membrane-bound COMT respectively). Increases bioavailability of levodopa when given as an adjunct therapy for Parkinson's disease. Inhibits α-synuclein aggregation in an in vitro assay; blocks α-synuclein-induced cell death in PC-12 cells.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 305.29 |
Percent Purity | >99% |
Chemical Name or Material | Entacapone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction